Anti-EPCAM Mouse Monoclonal Antibody [clone: OTI3G2]

Supplier: OriGene
TA506695
ORIGTA506695EA 616 EUR
ORIGTA506695
Anti-EPCAM Mouse Monoclonal Antibody [clone: OTI3G2]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: EPCAM (epithelial cell adhesion molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR