Anti-MMP13 Mouse Monoclonal Antibody [clone: OTI3F5]

Supplier: OriGene
TA506721
ORIGTA506721EA 616 EUR
ORIGTA506721
Anti-MMP13 Mouse Monoclonal Antibody [clone: OTI3F5]
Antibodies
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: MMP13 (matrix metallopeptidase 13 (collagenase 3))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR