Anti-UPRT Mouse Monoclonal Antibody [clone: OTI1C9]

Supplier: OriGene
ORIGTA506869EA 616 EUR
ORIGTA506869
Anti-UPRT Mouse Monoclonal Antibody [clone: OTI1C9]
Antibodies
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: UPRT (uracil phosphoribosyltransferase (FUR1) homolog (S. cerevisiae))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR