Anti-MFAP3 Mouse Monoclonal Antibody [clone: OTI1G1]

Supplier: OriGene
TA506943
ORIGTA506943EA 593 EUR
ORIGTA506943
Anti-MFAP3 Mouse Monoclonal Antibody [clone: OTI1G1]
Antibodies
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: MFAP3 (microfibrillar-associated protein 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR