Anti-TMX1 Mouse Monoclonal Antibody [clone: OTI1A4]

Supplier: OriGene
TA506947
ORIGTA506947EA 616 EUR
ORIGTA506947
Anti-TMX1 Mouse Monoclonal Antibody [clone: OTI1A4]
Antibodies
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: TMX1 (thioredoxin-related transmembrane protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR