Anti-SERPINB3 Mouse Monoclonal Antibody [clone: OTI2B9]

Supplier: OriGene
TA506997
ORIGTA506997EA 616 EUR
ORIGTA506997
Anti-SERPINB3 Mouse Monoclonal Antibody [clone: OTI2B9]
Antibodies
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: SERPINB3 (serpin peptidase inhibitor, clade B (ovalbumin), member 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR