Anti-HAUS7 Mouse Monoclonal Antibody [clone: OTI2F2]

Supplier: OriGene
TA507053
ORIGTA507053EA 616 EUR
ORIGTA507053
Anti-HAUS7 Mouse Monoclonal Antibody [clone: OTI2F2]
Antibodies
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: HAUS7 (HAUS augmin-like complex, subunit 7)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR