Anti-IL4 Mouse Monoclonal Antibody [clone: OTI8D4]

Supplier: OriGene
TA600022
ORIGTA600022EA 616 EUR
ORIGTA600022
Anti-IL4 Mouse Monoclonal Antibody [clone: OTI8D4]
Antibodies
Recommended Dilutions: 1:100-1:1000

Type: Primary
Antigen: IL4 (interleukin 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR