Anti-PRKCE Mouse Monoclonal Antibody [clone: 3F10]

Supplier: OriGene
TA600074
ORIGTA600074EA 616 EUR
ORIGTA600074
Anti-PRKCE Mouse Monoclonal Antibody [clone: 3F10]
Antibodies
Recommended Dilutions: 1:100-1:1000

Type: Primary
Antigen: PRKCE (protein kinase C, epsilon)
Clonality: Monoclonal
Clone: 3F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR