Anti-MGLL Mouse Monoclonal Antibody [clone: OTI2G8]

Supplier: OriGene
TA600158
ORIGTA600158EA 616 EUR
ORIGTA600158
Anti-MGLL Mouse Monoclonal Antibody [clone: OTI2G8]
Antibodies
Recommended Dilutions: 1:100-1:1000

Type: Primary
Antigen: MGLL (monoglyceride lipase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR