Anti-MYBL1 Mouse Monoclonal Antibody [clone: 2A2]

Supplier: Abnova
ABNOH00004603-M01EA 394 EUR
ABNOH00004603-M01
Anti-MYBL1 Mouse Monoclonal Antibody [clone: 2A2]
Antibodies
Type: Primary
Antigen: MYBL1
Clonality: Monoclonal
Clone: 2A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR