Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NPC2 Mouse Monoclonal Antibody [clone: 4B9]

Supplier: Abnova
Type: Primary
Antigen: NPC2
Clonality: Monoclonal
Clone: 4B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human



 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µg 100 µg ABNOH00010577-M01
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol NPC2
Antigen name Niemann-Pick disease type C2
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 4B9
Host Mouse
Isotype none
Western blot Yes
ELISA Yes
Antigen synonyms NPC2, epididymal secretory protein, MGC1333, epididymal protein 1, Niemann-Pick disease, EDDM1, NP-C2, type C2, HE1, tissue-specific secretory protein, epididymal secretory protein E1
Gene ID 10577
Cross adsorption non
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping