Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MXD4 Mouse Polyclonal Antibody

Supplier: Abnova
Type: Primary
Antigen: MXD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human



 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
50 µl 50 µl ABNOH00010608-B02
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol MXD4
Antigen name MAX dimerization protein 4
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Mouse
Isotype none
Western blot Yes
Antigen synonyms MSTP149, MST149, MAD4, bHLHc12
Gene ID 10608
Cross adsorption non
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty