Anti-D2s Dopamine Receptor Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB26713-100EA 0 EUR
BIRBORB26713-100
Anti-D2s Dopamine Receptor Rabbit Polyclonal Antibody
Antibodies
Immunochemistry: 1:500; Western Blot: 1:800

Type: Primary
Antigen: D2s Dopamine Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR