Anti-Nicotinic b2 Rec. Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB26735-100EA 0 EUR
BIRBORB26735-100
Anti-Nicotinic b2 Rec. Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Nicotinic b2 Rec.
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR