Anti-APP Mouse monoclonal antibody

Supplier: Biorbyt
BIRBORB26760-100EA 0 EUR
BIRBORB26760-100
Anti-APP Mouse monoclonal antibody
Antibodies
DB: 1 ?g/ml; ELISA: 0.1 ?g/ml

Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR