Anti-S-100 (beta-beta) Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB26769-1EA 0 EUR
BIRBORB26769-1
Anti-S-100 (beta-beta) Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: S-100 (beta-beta)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR