Anti-COL1 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB26827-0.2EA 0 EUR
BIRBORB26827-0.2
Anti-COL1 Goat Polyclonal Antibody
Antibodies
Immunochemistry: 1:10-1:20

Type: Primary
Antigen: COL1 (collagen, type I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR