Anti-Cytokeratin 8/18 Mouse monoclonal antibody

Supplier: Biorbyt
BIRBORB27210-5EA 0 EUR
BIRBORB27210-5
Anti-Cytokeratin 8/18 Mouse monoclonal antibody
Antibodies
Type: Primary
Antigen: Cytokeratin 8/18
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR