Anti-AGE Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB27266-0.2EA 0 EUR
BIRBORB27266-0.2
Anti-AGE Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:5000 - 1:25000

Type: Primary
Antigen: AGE
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR