Anti-DEFB103A Rabbit polyclonal antibody

Supplier: Biorbyt
BIRBORB27278-100EA 0 EUR
BIRBORB27278-100
Anti-DEFB103A Rabbit polyclonal antibody
Antibodies
Western Blot: 1:1000 - 1:5000; ELISA: 1:10000 - 1:50000

Type: Primary
Antigen: DEFB103A (defensin, beta 103A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR