Anti-AGE-3 Glycolaldehyde modified Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB27286-10EA 0 EUR
BIRBORB27286-10
Anti-AGE-3 Glycolaldehyde modified Mouse Monoclonal Antibody
Antibodies
Western Blot: 1 ?g/ml; ELISA: 0.1 ?g/ml

Type: Primary
Antigen: AGE-3 Glycolaldehyde modified
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR