Anti-COL3 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB27315-0.1EA 0 EUR
BIRBORB27315-0.1 BIRBORB27315-0.5
Anti-COL3 Rabbit Polyclonal Antibody
Antibodies
ImmunofluorescenceA 1:80; Immunochemistry: 1:500; ELISA: 1:200

Type: Primary
Antigen: COL3 (collagen, type III)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR