Anti-COL4 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB27322-0.1 ORB27322-0.5
BIRBORB27322-0.1EA 0 EUR
BIRBORB27322-0.1 BIRBORB27322-0.5
Anti-COL4 Rabbit Polyclonal Antibody
Antibodies
ImmunofluorescenceA 1:80; Immunochemistry: 1:500; ELISA: >1:200

Type: Primary
Antigen: COL4 (collagen, type IV)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR