Anti-ICAM1 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB27345-100EA 0 EUR
BIRBORB27345-100
Anti-ICAM1 Mouse Monoclonal Antibody
Antibodies
  • Immunochemistry: 2 μg/ml

Type: Primary
Antigen: ICAM1 (intercellular adhesion molecule 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR