Anti-CD34 Mouse Monoclonal Antibody

Supplier: Biorbyt
ORB27399-5
BIRBORB27399-5EA 0 EUR
BIRBORB27399-5
Anti-CD34 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CD34 (CD34 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR