Anti-Ig Lambda Chain Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB27419-5EA 0 EUR
BIRBORB27419-5
Anti-Ig Lambda Chain Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Ig Lambda Chain
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR