Anti-Melanoma Mouse Monoclonal Antibody

Supplier: Biorbyt
ORB27429-1
BIRBORB27429-1EA 0 EUR
BIRBORB27429-1
Anti-Melanoma Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Melanoma
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR