Anti-S100B Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB27461-1EA 0 EUR
BIRBORB27461-1
Anti-S100B Mouse Monoclonal Antibody
Antibodies
Immunochemistry: 1:20 - 1:50

Type: Primary
Antigen: S100B (S100 calcium binding protein B)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR