Anti-EDN1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB27485-50EA 0 EUR
BIRBORB27485-50
Anti-EDN1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EDN1 (endothelin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR