Anti-CML (N-Epsilon)-Carboxymethyl-Lysine Mouse Monoclonal Antibody

Supplier: Biorbyt
ORB27499-50
BIRBORB27499-50EA 0 EUR
BIRBORB27499-50
Anti-CML (N-Epsilon)-Carboxymethyl-Lysine Mouse Monoclonal Antibody
Antibodies
ELISA: 0.1-0.5 ?g/ml; Immunochemistry: 2 ?g/ml

Type: Primary
Antigen: CML (N-Epsilon)-Carboxymethyl-Lysine
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR