Anti-APP Rabbit polyclonal antibody

Supplier: Biorbyt
BIRBORB27575-100EA 0 EUR
BIRBORB27575-100
Anti-APP Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR