Anti-CD80 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB27578-100
BIRBORB27578-100EA 0 EUR
BIRBORB27578-100
Anti-CD80 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD80 (CD80 molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR