Anti-SYNPO Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB27596-100
BIRBORB27596-100EA 0 EUR
BIRBORB27596-100
Anti-SYNPO Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SYNPO (synaptopodin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR