Anti-GADD45A None polyclonal antibody

Supplier: Biorbyt
BIRBORB27612-100EA 0 EUR
BIRBORB27612-100
Anti-GADD45A None polyclonal antibody
Antibodies
Type: Primary
Antigen: GADD45A (growth arrest and DNA-damage-inducible, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: IgG (Rabbit)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR