Anti-XCL1 None Polyclonal Antibody

Supplier: Biorbyt
BIRBORB27616-100EA 0 EUR
BIRBORB27616-100
Anti-XCL1 None Polyclonal Antibody
Antibodies
Type: Primary
Antigen: XCL1 (chemokine (C motif) ligand 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: IgG (Rabbit)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR