Anti-PDPN None Polyclonal Antibody

Supplier: Biorbyt
BIRBORB27620-100EA 0 EUR
BIRBORB27620-100
Anti-PDPN None Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PDPN (podoplanin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR