Anti-IL12B Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB27649-100EA 0 EUR
BIRBORB27649-100
Anti-IL12B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL12B (interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR