Anti-FGF9 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB27671-100
BIRBORB27671-100EA 0 EUR
BIRBORB27671-100
Anti-FGF9 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FGF9 (fibroblast growth factor 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR