Anti-IgG Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB27721-0.5EA 0 EUR
BIRBORB27721-0.5 BIRBORB27721-1
Anti-IgG Goat Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR