Anti-IgG Rabbit Antibody

Supplier: Biorbyt
BIRBORB27746-5EA 0 EUR
BIRBORB27746-5 BIRBORB27746-1
Anti-IgG Rabbit Antibody
Antibodies
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR