Anti-Retinol binding protein 4 Mouse Monoclonal Antibody

Supplier: Biorbyt
ORB27758-100
BIRBORB27758-100EA 0 EUR
BIRBORB27758-100
Anti-Retinol binding protein 4 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Retinol binding protein 4
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR