Anti-APPL2 Rabbit polyclonal antibody

Supplier: Biorbyt
BIRBORB27764-100EA 0 EUR
BIRBORB27764-100
Anti-APPL2 Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: APPL2 (adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR