Anti-Plasminogen activator inhibitor type-1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB27775-100EA 0 EUR
BIRBORB27775-100
Anti-Plasminogen activator inhibitor type-1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Plasminogen activator inhibitor type-1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR