Anti-FGF21 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB27779-100EA 0 EUR
BIRBORB27779-100
Anti-FGF21 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FGF21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR