Anti-Histamine H3 Receptor Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB40788-100
BIRBORB40788-100EA 0 EUR
BIRBORB40788-100
Anti-Histamine H3 Receptor Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:3000

Type: Primary
Antigen: Histamine H3 Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR