Anti-RASSF1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB40798-100EA 0 EUR
BIRBORB40798-100
Anti-RASSF1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: RASSF1 (Ras association (RalGDS/AF-6) domain family member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR