Anti-CSE1L Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB40956-100
BIRBORB40956-100EA 0 EUR
BIRBORB40956-100
Anti-CSE1L Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:500; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: CSE1L (CSE1 chromosome segregation 1-like (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR