Anti-PPP1CC Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB41166-100EA 0 EUR
BIRBORB41166-100
Anti-PPP1CC Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: PPP1CC (protein phosphatase 1, catalytic subunit, gamma isozyme)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR