Anti-XRCC4 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB41555-100
BIRBORB41555-100EA 0 EUR
BIRBORB41555-100
Anti-XRCC4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: XRCC4 (X-ray repair complementing defective repair in Chinese hamster cells 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR